This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP786 SIRNA Assays ›
Gene Symbol:
Zfp786
Gene Name:
zinc finger protein 786
Gene Aliases:
A730012O14Rik, Znf786
Chromosome Location:
Chr.6: 47819263 - 47835920 on Build GRCm38
UniGene ID:
Mm.128149
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 99231

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp786
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177882.4 NP_808550.1 2 193
GenBank mRNA AK080438.1 - 2 193
BC138897.1 - 2 134 8861919

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001127594, XM_006236413, XM_006236414, BC166479.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleic acid binding
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0