This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CUL7 SIRNA Assays ›
Gene Symbol:
Cul7
Gene Name:
cullin 7
Gene Aliases:
2510004L20Rik, AA409809, C230011P08Rik, p185, p193
Chromosome Location:
Chr.17: 46650338 - 46664364 on Build GRCm38
UniGene ID:
Mm.329078
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 98025

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cul7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025611.5 NP_079887.3 5 1675
GenBank mRNA AK082134.1 - 1 3471
AK147156.1 - 5 1433
AK147905.1 - 5 1391
AK155454.1 - 4 2162
AK155583.1 - 5 1679
AK166135.1 - 4 2002
AK170580.1 - 5 1678
AK170808.1 - 5 1449
AK172882.1 - 4 748
BC059865.1 - 2 395
BC065096.1 - 4 1311
BC138998.1 - 5 1613 8862020
BC139000.1 - 5 1613 8862022

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
mitotic cytokinesis
vasculogenesis
epithelial to mesenchymal transition
placenta development
ubiquitin-dependent protein catabolic process
apoptotic process
Golgi organization
regulation of mitotic nuclear division
protein ubiquitination
regulation of apoptotic process
regulation of endothelial cell differentiation
positive regulation of dendrite morphogenesis
protein binding
ubiquitin protein ligase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-9hb2s:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header