This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM251 SIRNA Assays ›
Gene Symbol:
Tmem251
Gene Name:
transmembrane protein 251
Gene Aliases:
D230037D09Rik
Chromosome Location:
Chr.12: 102743687 - 102745406 on Build GRCm38
UniGene ID:
Mm.86546
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 97981

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tmem251
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177140.3 NP_796114.1 2 1183
GenBank mRNA AK052027.1 - 2 1185
AK144149.1 - 1185
AK172131.1 - 2 809
AK212553.1 - 1 296
AK219426.1 - 1 361

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_002729747.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0