This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4930412M03RIK SIRNA Assays ›
Gene Symbol:
4930412M03Rik
Gene Name:
RIKEN cDNA 4930412M03 gene
Gene Aliases:
-
Chromosome Location:
-
UniGene ID:
Mm.72458
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 97665

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4930412M03Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK029606.1 - 2 821
AK089395.1 - 1 2741
AK154788.1 - 1 2739
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA NM_177098.1 - 821 Chr. - - [-] on Build

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm