This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RELT SIRNA Assays ›
Gene Symbol:
Relt
Gene Name:
RELT tumor necrosis factor receptor
Gene Aliases:
E430021K24Rik, Tnfrsf19l
Chromosome Location:
Chr.7: 100845847 - 100863480 on Build GRCm38
UniGene ID:
Mm.40336
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 97548

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Relt
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177073.6 NP_796047.2 4 280
GenBank mRNA AK042363.1 - 4 275
AK049073.1 - 4 281
AK088621.1 - 4 263
BC100310.1 - 4 270 30839974
BC131929.1 - 4 191 40130206
BC131931.1 - 4 191 40130208
BC145200.1 - 4 191 9053751

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

programmed cell death
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0