This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CNPY1 SIRNA Assays ›
Gene Symbol:
Cnpy1
Gene Name:
canopy 1 homolog (zebrafish)
Gene Aliases:
1500012D20Rik, 9630008K15Rik, AI853839
Chromosome Location:
Chr.5: 28200826 - 28237873 on Build GRCm38
UniGene ID:
Mm.381479
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 96989

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cnpy1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175651.4 NP_783582.1 2 292
GenBank mRNA AK035828.1 - 2 293
AK035938.1 - 1 832
AK139155.1 - 1 978
AK162570.1 - 1 781
BC096464.1 - 217 6390940

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0