This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MLEC SIRNA Assays ›
Gene Symbol:
Mlec
Gene Name:
malectin
Gene Aliases:
2410014A08Rik, AI850530, ESTM19, X83352, mKIAA0152
Chromosome Location:
Chr.5: 115142981 - 115158176 on Build GRCm38
UniGene ID:
Mm.153963
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 96647

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mlec
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175403.3 NP_780612.2 5 1885
GenBank mRNA AK031525.1 - 5 1859
AK129066.1 - 5 1784

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
enzyme binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0