This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CHST8 SIRNA Assays ›
Gene Symbol:
Chst8
Gene Name:
carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 8
Gene Aliases:
1500011J21Rik, AI426009
Chromosome Location:
Chr.7: 34674468 - 34812711 on Build GRCm38
UniGene ID:
Mm.29380
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 96264

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Chst8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175140.4 NP_780349.3 4 2080
GenBank mRNA AB106878.1 - 3 1678
AK005217.1 - 4 1814
AK038952.1 - 3 2051
AK051290.1 - 4 1662
BC051124.1 - 4 2045 6528978

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
sulfur compound metabolic process
carbohydrate biosynthetic process
peptide hormone processing
proteoglycan biosynthetic process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0