This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SRRM2 SIRNA Assays ›
Gene Symbol:
Srrm2
Gene Name:
serine/arginine repetitive matrix 2
Gene Aliases:
5033413A03Rik, AA410130, SRm300, mKIAA0324
Chromosome Location:
Chr.17: 23790857 - 23824741 on Build GRCm38
UniGene ID:
Mm.436876
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 96244

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Srrm2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175229.3 NP_780438.2 5, 6 634
GenBank mRNA AK090123.1 - 5, 6 595
AK171001.1 - 6, 7 907

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016333, AB002322, BX647088, AB015644, AB016087, and others.
This siRNA also targets the following transcripts in Rat: NM_001277154, FQ234799.

Gene Ontology Categories:

Function(s)

mRNA processing
biological_process
RNA splicing

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0