This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MVB12B SIRNA Assays ›
Gene Symbol:
Mvb12b
Gene Name:
multivesicular body subunit 12B
Gene Aliases:
2610200O14Rik, 2610528K11Rik, AI414895, Fam125b
Chromosome Location:
Chr.2: 33729953 - 33887946 on Build GRCm38
UniGene ID:
Mm.254914
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 96206

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Mvb12b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175184.4 NP_780393.2 3 821
GenBank mRNA AK012178.1 - 3 314
AK087821.1 - 3 339
AK131113.1 - 3 339
AK135990.1 - 3 255
AK154267.1 - 3 315
BC049129.1 - 3 334 6825067
BC059907.1 - 3 821 6824889

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_033446, NM_001011703, AK024432, AK298023, BC028675, and others.
This siRNA also targets the following transcripts in Rat: NM_001271232, XM_008761761.

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
virus maturation
regulation of epidermal growth factor receptor signaling pathway
positive regulation of viral process
lipid binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header