This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GOLIM4 SIRNA Assays ›
Gene Symbol:
Golim4
Gene Name:
golgi integral membrane protein 4
Gene Aliases:
3110027H23Rik, GIMPC, GPP130, Golph4, P138
Chromosome Location:
Chr.3: 75876183 - 75956949 on Build GRCm38
UniGene ID:
Mm.249232
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 96118

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Golim4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291069.1 NP_001277998.1 2 895
NM_175193.6 NP_780402.1 2 895
GenBank mRNA AK033730.1 - 2 882
AK048434.1 - 3 806
AK078928.1 - 2 895
AK081156.1 - 2 599
AK149286.1 - 2 855
BC086642.1 - 2 727 6836803
BC089354.1 - 2 777 6837211

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001191567, XM_006232499, XM_006232500.

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0