This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ATAD3AOS SIRNA Assays ›
Gene Symbol:
Atad3aos
Gene Name:
ATPase family, AAA domain containing 3A, opposite strand
Gene Aliases:
2610204G22Rik
Chromosome Location:
Chr.4: 155761192 - 155763560 on Build GRCm38
UniGene ID:
Mm.70636
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 96088

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Atad3aos
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK078249.1 - 2 671

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0