This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other POU6F2 SIRNA Assays ›
Gene Symbol:
Pou6f2
Gene Name:
POU domain, class 6, transcription factor 2
Gene Aliases:
D130006K24Rik, RPF-1
Chromosome Location:
Chr.13: 18124960 - 18382044 on Build GRCm38
UniGene ID:
Mm.352263
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95848

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pou6f2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175006.2 NP_778171.2 205
GenBank mRNA AK083772.1 - 205
BC094915.1 - 220 6435959
BC138151.1 - 124 8861170

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_007252, NM_001166018, DN991890, AK309700, U91935.
This siRNA also targets the following transcripts in Rat: NM_001101002, BC128774.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0