This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VPS13A SIRNA Assays ›
Gene Symbol:
Vps13a
Gene Name:
vacuolar protein sorting 13A (yeast)
Gene Aliases:
4930425F11, 4930516E05Rik, 4930543C13Rik, 9930023P20, Chac, Chorein, D330038K10Rik, mKIAA0986
Chromosome Location:
Chr.19: 16612194 - 16781139 on Build GRCm38
UniGene ID:
Mm.211963
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95408

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vps13a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173028.4 NP_766616.2 49 7010
GenBank mRNA AB115421.1 - 49 7005
AK029649.1 - 3 765
AK030073.1 - 2 234
AK135983.1 - 6 1164
AK220362.1 - 2 427
BC062130.1 - 5 908
BC145012.1 - 49 6899

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein targeting to vacuole
transport
nervous system development
locomotory behavior
protein transport
social behavior
protein retention in Golgi apparatus
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header