This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ST6GAL2 SIRNA Assays ›
Gene Symbol:
St6gal2
Gene Name:
beta galactoside alpha 2,6 sialyltransferase 2
Gene Aliases:
C230064G14Rik, St6galII, mKIAA1877
Chromosome Location:
Chr.17: 55444358 - 55514581 on Build GRCm38
UniGene ID:
Mm.311687
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 95061

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: St6gal2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172829.2 NP_766417.1 2 811
GenBank mRNA AK054230.1 - 2 1147
AK082566.1 - 2 812
BC125586.1 - 1 681 40130101
BC138142.1 - 1 681 8861161

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
N-acetylneuraminate metabolic process
protein glycosylation
oligosaccharide metabolic process
sialylation
catalytic activity
beta-galactoside alpha-2,6-sialyltransferase activity
sialyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0