This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CLEC9A SIRNA Assays ›
Gene Symbol:
Clec9a
Gene Name:
C-type lectin domain family 9, member a
Gene Aliases:
9830005G06Rik, DNGR-1
Chromosome Location:
Chr.6: 129408862 - 129501701 on Build GRCm38
UniGene ID:
Mm.391518
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 94779

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Clec9a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001205363.1 NP_001192292.1 3 281
NM_001205364.1 NP_001192293.1 2 200
NM_001205365.1 NP_001192294.1 3 281
NM_172732.3 NP_766320.2 3 281
GenBank mRNA AK036399.1 - 3 282
AK041288.1 - 3 281
BC087955.1 - 3 280 30119997
EU339277.1 - 3 188
EU339278.1 - 3 188
EU339279.1 - 2 107
EU339280.1 - 3 188
EU339281.1 - 3 188

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
positive regulation of cytokine secretion
carbohydrate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0