This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLHL4 SIRNA Assays ›
Gene Symbol:
Klhl4
Gene Name:
kelch-like 4
Gene Aliases:
B130014A11, C130018J01Rik
Chromosome Location:
Chr.X: 114474333 - 114561129 on Build GRCm38
UniGene ID:
Mm.132583
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 94729

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Klhl4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290477.1 NP_001277406.1 3 762
NM_001290478.1 NP_001277407.1 3 762
NM_172781.2 NP_766369.2 3 762
GenBank mRNA AK044930.1 - 3 493
AK081448.1 - 3 763
AK081937.1 - 3 575
AK085099.1 - 3 763
AK161786.1 - 3 656
BC057137.1 - 3 493
BC099954.1 - 3 589 30081872
BC141251.1 - 3 505 9055863

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0