This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other APBB1 SIRNA Assays ›
Gene Symbol:
APBB1
Gene Name:
amyloid beta precursor protein binding family B member 1
Gene Aliases:
FE65, MGC:9072, RIR
Chromosome Location:
Chr.11: 6395124 - 6419830 on Build GRCh38
UniGene ID:
Hs.372840
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 9454

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: APBB1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164.4 NP_001155.1 2 483
NM_145689.2 NP_663722.1 2 483
NR_047512.2 2 483
XM_006718209.3 XP_006718272.2 2 518
XM_006718210.3 XP_006718273.2 2 568
XM_006718211.1 XP_006718274.1 2 724
XM_006718212.3 XP_006718275.2 2 518
XM_006718213.1 XP_006718276.1 2 724
XM_006718214.3 XP_006718277.2 2 518
XM_006718215.3 XP_006718278.3 2 841
XM_017017639.1 XP_016873128.1 2 724
XM_017017640.1 XP_016873129.1 2 724
XM_017017641.1 XP_016873130.1 2 724
GenBank mRNA AF394214.1 - 2 442
BC010854.2 - 2 437 3905603
BG333769.1 - 2 478
BG699532.1 - 2 483
L77864.1 - 2 442

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
apoptotic process
cellular response to DNA damage stimulus
cell cycle arrest
signal transduction
axonogenesis
response to iron ion
positive regulation of neuron projection development
negative regulation of cell growth
positive regulation of apoptotic process
histone H4 acetylation
positive regulation of DNA repair
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein secretion
negative regulation of thymidylate synthase biosynthetic process
beta-amyloid binding
chromatin binding
protein binding
transcription factor binding
protein complex binding
histone binding
tau protein binding
proline-rich region binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0