This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TDRD3 SIRNA Assays ›
Gene Symbol:
Tdrd3
Gene Name:
tudor domain containing 3
Gene Aliases:
4732418C03Rik, 6720468N18
Chromosome Location:
Chr.14: 87416583 - 87545508 on Build GRCm38
UniGene ID:
Mm.35168
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 94480

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tdrd3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253755.1 NP_001240684.1 8 1028
NM_172605.3 NP_766193.3 8 1028
GenBank mRNA AK032892.1 - 8 973
AK033390.1 - 8 973
AK181483.1 - 2 223
BC005670.1 - 8 929 3153274
BC066144.1 - 8 976 30435963

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mRNA processing
chromatin modification
regulation of nucleic acid-templated transcription
chromatin binding
transcription coactivator activity
RNA binding
methylated histone binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0