This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NUTM1 SIRNA Assays ›
Gene Symbol:
Nutm1
Gene Name:
NUT midline carcinoma, family member 1
Gene Aliases:
4932438M10, Nut
Chromosome Location:
Chr.2: 112247948 - 112259291 on Build GRCm38
UniGene ID:
Mm.78965
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 94213

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nutm1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172521.1 NP_766109.1 2 662
GenBank mRNA AK077049.1 - 2 662
BC125332.1 - 2 642 40129847
BC125334.1 - 2 642 40129849

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108957, XM_006234708.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0