This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GABPB2 SIRNA Assays ›
Gene Symbol:
Gabpb2
Gene Name:
GA repeat binding protein, beta 2
Gene Aliases:
1810015F01Rik, 5830427M07, 9430006E19Rik, A430024B14Rik, AV050852, Gabpb2-1
Chromosome Location:
Chr.3: 95181766 - 95217951 on Build GRCm38
UniGene ID:
Mm.224820
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 94204

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gabpb2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029885.1 NP_084161.1 6, 7 1017
NM_172512.2 NP_766100.1 5, 6 920
GenBank mRNA AK030849.1 - 5, 6 689
AK036692.1 - 5, 6 1501
AK141382.1 - 6, 7 991
AK163939.1 - 5, 6 1497
BC150160.1 - 6, 7 768 40141577
BC150482.1 - 6, 7 768 40141578

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_144618, AK313606, AK092230, AK301882, BC027033.
This siRNA also targets the following transcripts in Rat: XM_227439, XM_006232936.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein homodimerization activity
transcription regulatory region DNA binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0