This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM126B SIRNA Assays ›
Gene Symbol:
Fam126b
Gene Name:
family with sequence similarity 126, member B
Gene Aliases:
C130065N10Rik, D1Ertd53e, D630010C10
Chromosome Location:
Chr.1: 58522806 - 58586333 on Build GRCm38
UniGene ID:
Mm.26272
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 94109

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam126b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172513.3 NP_766101.3 4, 5 396
GenBank mRNA AK052638.1 - 4, 5 323
AK144602.1 - 4, 5 333
AK168372.1 - 4, 5 386
BC049806.1 - 4, 5 333
BC068302.1 - 4, 5 304 30431671

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0