This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DPYD SIRNA Assays ›
Gene Symbol:
Dpyd
Gene Name:
dihydropyrimidine dehydrogenase
Gene Aliases:
AI315208, DPD, E330028L06Rik
Chromosome Location:
Chr.3: 118562109 - 119432918 on Build GRCm38
UniGene ID:
Mm.27907
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 93511

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dpyd
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_170778.2 NP_740748.1 23 3696
GenBank mRNA AK054474.1 - 4 1122
BC028831.1 - 5 1240
BC039699.1 - 23 3697 4235971
BC042543.1 - 19 3161
BC044730.2 - 11 2059

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

purine nucleobase catabolic process
pyrimidine nucleobase catabolic process
thymine catabolic process
uracil catabolic process
thymidine catabolic process
UMP biosynthetic process
response to nutrient
circadian rhythm
uracil metabolic process
response to drug
oxidation-reduction process
nucleotide binding
uracil binding
catalytic activity
dihydroorotate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
dihydropyrimidine dehydrogenase (NADP+) activity
protein homodimerization activity
metal ion binding
NADP binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0