This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM210A SIRNA Assays ›
Gene Symbol:
Fam210a
Gene Name:
family with sequence similarity 210, member A
Gene Aliases:
4933403F05Rik, AA987140, AV134576, AW491926
Chromosome Location:
Chr.18: 68260184 - 68300333 on Build GRCm38
UniGene ID:
Mm.125857
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 93386

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam210a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153794.4 NP_722489.1 2 405
GenBank mRNA AK034980.1 - 2 287
AK044238.1 - 2 293
AK049567.1 - 2 293
AK077093.1 - 2 406
AK142190.1 - 2 277
AK147662.1 - 2 280
AK159319.1 - 2 256
AK162396.1 - 2 290
BC038695.1 - 2 172 1548421
BC095936.1 - 2 255 3968496

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0