This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM210A SIRNA Assays ›
Gene Symbol:
Fam210a
Gene Name:
family with sequence similarity 210, member A
Gene Aliases:
4933403F05Rik, AA987140, AV134576, AW491926
Chromosome Location:
Chr.18: 68260184 - 68300333 on Build GRCm38
UniGene ID:
Mm.125857
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 93292

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam210a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153794.4 NP_722489.1 2 376
GenBank mRNA AK034980.1 - 2 258
AK044238.1 - 2 264
AK049567.1 - 2 264
AK077093.1 - 2 377
AK142190.1 - 2 248
AK147662.1 - 2 251
AK159319.1 - 2 227
AK162396.1 - 2 261
BC038695.1 - 2 143 1548421
BC095936.1 - 2 226 3968496

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header