This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZFP740 SIRNA Assays ›
Gene Symbol:
Zfp740
Gene Name:
zinc finger protein 740
Gene Aliases:
1110034O07Rik, AW548228, Znf740
Chromosome Location:
Chr.15: 102203645 - 102215610 on Build GRCm38
UniGene ID:
Mm.352945
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 92854

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Zfp740
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289690.1 NP_001276619.1 6 1116
NM_001289691.1 NP_001276620.1 8 1315
NM_001289694.1 NP_001276623.1 7 1024
NM_001289695.1 NP_001276624.1 7 990
GenBank mRNA AK144707.1 - 8 1312
AK170207.1 - 7 1012
BC030410.1 - 3 821 5367159
BC066011.1 - 7 884 6418561

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
nucleic acid binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0