This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDC5L SIRNA Assays ›
Gene Symbol:
Cdc5l
Gene Name:
cell division cycle 5-like (S. pombe)
Gene Aliases:
1200002I02Rik, AA408004, PCDC5RP
Chromosome Location:
Chr.17: 45391887 - 45433707 on Build GRCm38
UniGene ID:
Mm.28270
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 92628

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdc5l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152810.2 NP_690023.1 13 2078
GenBank mRNA AK004547.2 - 13 2108
AK146892.1 - 13 1898
AK150562.1 - 13 1895
AK150861.1 - 13 1895
AK172950.1 - 13 1898
BC031480.1 - 13 2078 3498853

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
mRNA processing
cellular response to DNA damage stimulus
cell cycle
RNA splicing
signal transduction involved in DNA damage checkpoint
DNA binding
chromatin binding
RNA binding
protein domain specific binding
poly(A) RNA binding
WD40-repeat domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0