siRNA Details
Gene Symbol: Olfr905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146804.2 | NP_667015.2 | 1 | 819 | ||
GenBank mRNA | BC150476.1 | - | 1 | 823 | 40134740 |

Gene Symbol: Olfr905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146804.2 | NP_667015.2 | 1 | 819 | ||
GenBank mRNA | BC150476.1 | - | 1 | 823 | 40134740 |