This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other OLFR1341 SIRNA Assays ›
Gene Symbol:
Olfr1341
Gene Name:
olfactory receptor 1341
Gene Aliases:
MOR258-1
Chromosome Location:
Chr.4: 118709409 - 118710347 on Build GRCm38
UniGene ID:
Mm.377720
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 91765

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Olfr1341
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146853.2 NP_667064.2 1 400
GenBank mRNA BC130239.1 - 1 404
BC152333.1 - 1 404
BC152353.1 - 1 404

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000584, XM_003750009.

Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
sensory perception of smell
detection of chemical stimulus involved in sensory perception of smell
olfactory receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0