siRNA Details
Gene Symbol: Olfr446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146295.1 | NP_666407.1 | 1 | 603 | ||
GenBank mRNA | BC087904.1 | - | 1077 | |||
BC104060.1 | - | 611 | ||||
BC104061.1 | - | 610 |

Gene Symbol: Olfr446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146295.1 | NP_666407.1 | 1 | 603 | ||
GenBank mRNA | BC087904.1 | - | 1077 | |||
BC104060.1 | - | 611 | ||||
BC104061.1 | - | 610 |