This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C8A SIRNA Assays ›
Gene Symbol:
C8a
Gene Name:
complement component 8, alpha polypeptide
Gene Aliases:
-
Chromosome Location:
Chr.4: 104815679 - 104876486 on Build GRCm38
UniGene ID:
Mm.197638
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 90718

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C8a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146148.2 NP_666260.1 2, 3 212
GenBank mRNA AB077295.1 - 2, 3 226
AB077296.1 - 2, 3 175
AB077298.1 - 2, 3 175
AK090245.1 - 2, 3 210
BC027748.1 - 2, 3 208 5052412

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

immune system process
immune response
complement activation
complement activation, alternative pathway
complement activation, classical pathway
cytolysis
innate immune response
complement binding
protein complex binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0