This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDCA7L SIRNA Assays ›
Gene Symbol:
Cdca7l
Gene Name:
cell division cycle associated 7 like
Gene Aliases:
BC006933, JPO2
Chromosome Location:
Chr.12: 117843475 - 117878712 on Build GRCm38
UniGene ID:
Mm.281149
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 90443

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdca7l
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146040.1 NP_666152.1 2 171
GenBank mRNA AK149673.1 - 2 149
BC006933.1 - 2 171 3602150

Back To Top

More Information


Gene Ontology Categories:

Function(s)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of cell proliferation

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0