This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM193B SIRNA Assays ›
Gene Symbol:
Fam193b
Gene Name:
family with sequence similarity 193, member B
Gene Aliases:
IRIZIO, Kiaa1931
Chromosome Location:
Chr.13: 55539316 - 55571120 on Build GRCm38
UniGene ID:
Mm.35661
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 89494

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam193b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145382.4 NP_663357.2 8 3318
GenBank mRNA AK040568.1 - 2 1446
AK155237.1 - 8 3312
AK173303.1 - 8 2167
BC021381.1 - 2 908 5043282
BC023015.1 - 2 850 5364758
BC025483.1 - 8 2177 5150988
BC051255.1 - 2 3766

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0