This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM20B SIRNA Assays ›
Gene Symbol:
Fam20b
Gene Name:
family with sequence similarity 20, member B
Gene Aliases:
C530043G21Rik, mKIAA0475
Chromosome Location:
Chr.1: 156678545 - 156719027 on Build GRCm38
UniGene ID:
Mm.229107
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 89426

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam20b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145413.4 NP_663388.1 2 489
GenBank mRNA AK034081.1 - 3 596
AK049705.1 - 1 291
AK132149.1 - 2 490
BC019381.1 - 2 508 4206622
BC023737.1 - 3 705 5352660
BC024412.1 - 1 439 4971483
BC031473.1 - 2 601 3990487

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

phosphorylation
positive regulation of protein targeting to mitochondrion
nucleotide binding
ATP binding
phosphofructokinase activity
phosphorylase kinase regulator activity
cobinamide kinase activity
phytol kinase activity
kinase activity
phosphatidylinositol phosphate kinase activity
cyclin-dependent protein serine/threonine kinase regulator activity
transferase activity
phosphotransferase activity, alcohol group as acceptor
phenol kinase activity
cyclin-dependent protein kinase activating kinase regulator activity
inositol tetrakisphosphate 2-kinase activity
heptose 7-phosphate kinase activity
aminoglycoside phosphotransferase activity
phosphatidylinositol 3-kinase activity
eukaryotic elongation factor-2 kinase regulator activity
eukaryotic elongation factor-2 kinase activator activity
LPPG:FO 2-phospho-L-lactate transferase activity
cytidine kinase activity
glycerate 2-kinase activity
(S)-lactate 2-kinase activity
phosphoserine:homoserine phosphotransferase activity
L-seryl-tRNA(Sec) kinase activity
phosphocholine transferase activity
polynucleotide 5'-hydroxyl-kinase activity
ATP-dependent polynucleotide kinase activity
GTP-dependent polynucleotide kinase activity
farnesol kinase activity
CTP:2-trans,-6-trans-farnesol kinase activity
geraniol kinase activity
geranylgeraniol kinase activity
CTP:geranylgeraniol kinase activity
prenol kinase activity
1-phosphatidylinositol-5-kinase activity
1-phosphatidylinositol-3-phosphate 4-kinase activity
phosphatidylinositol-3,4-bisphosphate 5-kinase activity
phosphatidylinositol bisphosphate kinase activity
inositol-3,4,6-trisphosphate 1-kinase activity
inositol 5-diphosphate pentakisphosphate 5-kinase activity
inositol diphosphate tetrakisphosphate kinase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-77574795ff-5hfml:80/100.66.128.142:80.
git-commit: 5cc8a939e1477954742eeb281a87e54f28adfb77
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header