This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other UGT3A2 SIRNA Assays ›
Gene Symbol:
Ugt3a2
Gene Name:
UDP glycosyltransferases 3 family, polypeptide A2
Gene Aliases:
AI313915
Chromosome Location:
Chr.15: 9335551 - 9370955 on Build GRCm38
UniGene ID:
Mm.422853
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 88938

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ugt3a2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144845.3 NP_659094.1 4 749
GenBank mRNA AK041045.1 - 4 734
AK050128.1 - 4 750
AK052644.1 - 4 759
AK143815.1 - 4 764
BC022134.1 - 4 734 5098754
BC034837.1 - 4 730 5134131

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
flavonoid biosynthetic process
cellular response to hormone stimulus
flavonoid glucuronidation
cellular response to genistein
UDP-glycosyltransferase activity
glucuronosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
transferase activity, transferring hexosyl groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0