This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other BC021614 SIRNA Assays ›
Gene Symbol:
BC021614
Gene Name:
cDNA sequence BC021614
Gene Aliases:
-
Chromosome Location:
Chr.19: 4057470 - 4059588 on Build GRCm38
UniGene ID:
Mm.440201
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 88767

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BC021614
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144869.2 NP_659118.1 2 111
GenBank mRNA BC021614.1 - 2 111 5102505
BC034269.1 - 3 124 4196879

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001134508, XM_006230736, XM_008760100, XM_006230737, FQ219091.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0