This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CDC26 SIRNA Assays ›
Gene Symbol:
Cdc26
Gene Name:
cell division cycle 26
Gene Aliases:
2010012C09Rik
Chromosome Location:
Chr.4: 62393812 - 62408640 on Build GRCm38
UniGene ID:
Mm.109930
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 88424

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cdc26
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139291.3 NP_647452.1 4 387
GenBank mRNA AK004518.1 - 1 28
AK012627.1 - 4 388
AK047055.1 - 407
AK086692.1 - 866
AK135556.1 - 387
BC005775.1 - 3 314 3602527
BC114350.2 - 2 174 40053418

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013240, NM_001267055, NM_001267054, XM_006238241, XM_008763796, and others.

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
regulation of mitotic metaphase/anaphase transition
anaphase-promoting complex-dependent catabolic process
cell division
protein K11-linked ubiquitination
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0