This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCTD9 SIRNA Assays ›
Gene Symbol:
Kctd9
Gene Name:
potassium channel tetramerisation domain containing 9
Gene Aliases:
-
Chromosome Location:
Chr.14: 67716098 - 67742310 on Build GRCm38
UniGene ID:
Mm.6720
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 87815

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kctd9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001111028.1 NP_001104498.1 12 1683
NM_001285933.1 NP_001272862.1 12 1810
NM_134073.2 NP_598834.2 11 1533
GenBank mRNA AK158922.1 - 11 1535
AK166501.1 - 11 1368
BC011302.1 - 12 1624

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein homooligomerization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header