This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ANO10 SIRNA Assays ›
Gene Symbol:
Ano10
Gene Name:
anoctamin 10
Gene Aliases:
AI604832, Tmem16k
Chromosome Location:
Chr.9: 122175874 - 122294443 on Build GRCm38
UniGene ID:
Mm.87676
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 87487

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ano10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271873.1 NP_001258802.1 2, 3 281
NM_133979.3 NP_598740.1 2, 3 281
GenBank mRNA AK043011.1 - 2, 3 244
AK046091.1 - 2, 3 233
AK079622.1 - 2, 3 232
AK080947.1 - 2, 3 233
AK138414.1 - 2, 3 267
BC099688.1 - 2, 3 204 6313494

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cation transport
chloride transport
cation transmembrane transport
chloride transmembrane transport
calcium activated cation channel activity
intracellular calcium activated chloride channel activity
phospholipid scramblase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header