This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other GPN2 SIRNA Assays ›
Gene Symbol:
Gpn2
Gene Name:
GPN-loop GTPase 2
Gene Aliases:
AI838661, Atpbd1b, R74630
Chromosome Location:
Chr.4: 133584373 - 133591735 on Build GRCm38
UniGene ID:
Mm.274001
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 87143

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gpn2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290742.1 NP_001277671.1 1 404
NM_133884.2 NP_598645.2 1 404
GenBank mRNA BC018407.1 - 1 397 4015340
CO798620.2 - 1 404

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001270959.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
molecular_function
GTP binding
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0