This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other 4931406C07RIK SIRNA Assays ›
Gene Symbol:
4931406C07Rik
Gene Name:
RIKEN cDNA 4931406C07 gene
Gene Aliases:
-
Chromosome Location:
Chr.9: 15283337 - 15306448 on Build GRCm38
UniGene ID:
Mm.261842
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 86860

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 4931406C07Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199484.1 NP_001186413.1 3 464
NM_001199485.1 NP_001186414.1 3 360
NM_133732.3 NP_598493.1 2 323
GenBank mRNA AK088123.1 - 3 324
AK133192.1 - 3 465
AK147073.1 - 3 334
BC011540.1 - 2 226 2648220
BC016078.1 - 2 272 4489072
BC025213.1 - 2 224
CT010296.1 - 2 137

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

metabolic process
nuclease activity
zinc ion binding
hydrolase activity
hydrolase activity, acting on ester bonds
phosphoric ester hydrolase activity
T/G mismatch-specific endonuclease activity
retroviral 3' processing activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0