This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TM9SF2 SIRNA Assays ›
Gene Symbol:
Tm9sf2
Gene Name:
transmembrane 9 superfamily member 2
Gene Aliases:
1500001N15Rik, AA536814, D14Ertd64e, P76
Chromosome Location:
Chr.14: 122107082 - 122159603 on Build GRCm38
UniGene ID:
Mm.275191
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 86439

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tm9sf2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080556.3 NP_542123.3 17 2610
GenBank mRNA AK010325.1 - 17 2566
AK030340.1 - 17 2541
AK048012.1 - 17 2529
AK050338.1 - 17 2557
AK075720.1 - 17 2612
AK088905.1 - 17 2582
AK159757.1 - 17 2556
AV028818.2 - 1 197
BC003862.1 - 17 2549 3488326
BC018595.1 - 1 450

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0