This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SMARCA5 SIRNA Assays ›
Gene Symbol:
Smarca5
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 5
Gene Aliases:
4933427E24Rik, D030040M08Rik, D330027N15Rik, MommeD4, Snf2h
Chromosome Location:
Chr.8: 80699943 - 80739459 on Build GRCm38
UniGene ID:
Mm.246803
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 85694

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Smarca5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053124.2 NP_444354.2 18 2746
GenBank mRNA AF325921.1 - 18 2398
AF375046.1 - 18 2465
AK052320.1 - 6 699
AK153789.1 - 18 3064
AK156915.1 - 2 210
AK157142.1 - 2757
BC021922.1 - 6 684
BC053069.1 - 18 2746 6409140

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chromatin silencing at rDNA
double-strand break repair
chromatin assembly or disassembly
nucleosome assembly
chromatin remodeling
DNA-templated transcription, initiation
embryo development
chromatin modification
nucleosome positioning
ATP-dependent chromatin remodeling
positive regulation of transcription, DNA-templated
nucleotide binding
nucleic acid binding
DNA binding
chromatin binding
helicase activity
protein binding
ATP binding
hydrolase activity
hydrolase activity, acting on acid anhydrides
hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
ATPase activity
nucleosome binding
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0