This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TRPS1 SIRNA Assays ›
Gene Symbol:
Trps1
Gene Name:
trichorhinophalangeal syndrome I (human)
Gene Aliases:
AI115454, AI447310, D15Ertd586e
Chromosome Location:
Chr.15: 50654757 - 50890747 on Build GRCm38
UniGene ID:
Mm.30466
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 85137

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Trps1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032000.2 NP_114389.2 2 521
GenBank mRNA AF346836.1 - 3 446
AK142355.1 - 2 191
BC037058.1 - 2 199 4951294
BC049857.1 - 3 225 4460789
BC099962.1 - 2 486 30091008

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chondrocyte differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
transmembrane receptor protein serine/threonine kinase signaling pathway
regulation of histone deacetylation
regulation of chondrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm