This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FAM131B SIRNA Assays ›
Gene Symbol:
Fam131b
Gene Name:
family with sequence similarity 131, member B
Gene Aliases:
6330503C03Rik, 6530406I18Rik, AV277466
Chromosome Location:
Chr.6: 42315305 - 42324640 on Build GRCm38
UniGene ID:
Mm.234314
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 83792

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fam131b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113327.2 NP_001106798.1 4 421
NM_001286584.1 NP_001273513.1 5 397
NM_029528.5 NP_083804.3 5 457
GenBank mRNA AK018206.1 - 5 396
AK018325.1 - 5 403
AK047763.1 - 5 383
AK158869.1 - 5 350
AK163654.1 - 4 342
BC115595.1 - 4 187
BC115596.1 - 4 187

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
mitophagy in response to mitochondrial depolarization
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0