This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC101 SIRNA Assays ›
Gene Symbol:
Ccdc101
Gene Name:
coiled-coil domain containing 101
Gene Aliases:
1700023O11Rik, 9530025I05Rik
Chromosome Location:
Chr.7: 126649309 - 126672779 on Build GRCm38
UniGene ID:
Mm.45686
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 83551

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ccdc101
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029339.3 NP_083615.3 6 503
GenBank mRNA AK006287.1 - 6 413
AK139102.1 - 6 504
AK169898.1 - 6 499
BC026784.1 - 6 441 4206090

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001114502, XM_006230232, XM_006230231, FQ210727, FQ232716.

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
histone H3 acetylation
enzyme binding
methylated histone binding
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header