This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TM9SF1 SIRNA Assays ›
Gene Symbol:
Tm9sf1
Gene Name:
transmembrane 9 superfamily member 1
Gene Aliases:
1200014D02Rik, AI893436, MP70
Chromosome Location:
Chr.14: 55635965 - 55643806 on Build GRCm38
UniGene ID:
Mm.132857
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 83193

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tm9sf1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028780.3 NP_083056.2 2 544
GenBank mRNA AF269152.1 - 2 192
AK004754.2 - 2 546
AK090065.1 - 1 539
AK152476.1 - 2 324
AK152540.1 - 2 324
AK160610.1 - 1 536
BC017617.1 - 1 483 4923337

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001012155, XM_006251991, XM_006251990, XM_008770688, BC081891.

Gene Ontology Categories:

Function(s) Process(es)

autophagy
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0