This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FUNDC1 SIRNA Assays ›
Gene Symbol:
Fundc1
Gene Name:
FUN14 domain containing 1
Gene Aliases:
1500005J14Rik, 1810033P05Rik
Chromosome Location:
Chr.X: 17519967 - 17572330 on Build GRCm38
UniGene ID:
Mm.41558
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 82583

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Fundc1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028058.3 NP_082334.1 5 650
GenBank mRNA AK005163.1 - 5 651
AK007686.1 - 4 687
AK168868.1 - 5 637
BC012515.1 - 5 592 2651125
BC030487.1 - 5 653 5374103

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitophagy
response to hypoxia
autophagy
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0