This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other STK11IP SIRNA Assays ›
Gene Symbol:
Stk11ip
Gene Name:
serine/threonine kinase 11 interacting protein
Gene Aliases:
1200014D22Rik, BB131189, LIP1, LKB1IP
Chromosome Location:
Chr.1: 75521529 - 75537335 on Build GRCm38
UniGene ID:
Mm.312650
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 82220

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Stk11ip
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027886.3 NP_082162.3 7, 8 754
GenBank mRNA AK004757.2 - 7, 8 707
AK156122.1 - 6, 7 842
AK165911.1 - 7, 8 745
AK171989.1 - 6, 7 616
BC042403.1 - 6, 7 613
BC061635.1 - 7, 8 737 6530178

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein localization
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0