This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other AVPI1 SIRNA Assays ›
Gene Symbol:
Avpi1
Gene Name:
arginine vasopressin-induced 1
Gene Aliases:
2310008N12Rik, AA410041, VIT32, esu, mVIT32
Chromosome Location:
Chr.19: 42123275 - 42128993 on Build GRCm38
UniGene ID:
Mm.30060
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 81614

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Avpi1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027106.4 NP_081382.1 3 623
GenBank mRNA AK009243.1 - 3 624
AK177091.1 - 1 115
BC027646.1 - 3 657 1344413

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
cell cycle
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0